Lindberg/Blue M Box Furnaces, 1200 °C, Thermo Scientific
Supplier: Thermo Fisher Scientific
BF51842C-1 BF51842PBC-1 BF51731BC-1
30604-200EA 19592 CAD
30604-200 30604-203 30712-122 30712-132
Lindberg/Blue M Box Furnaces, 1200 °C, Thermo Scientific
Ovens Furnaces (Ashing and Muffle Ovens)
Furnaces are designed for continuous operation to 1200 °C maximum and feature an integral controller in a single space-saving cabinet.
  • LGO heating elements and Moldatherm insulation
  • Variable heatup rate eliminates thermal shock to materials
  • Air vent (1" diameter, top) and air inlet (0.375" diameter, rear) for inert gas
  • Platinel II thermocouple for long-term stability
  • Removable shelves for versatility
  • Self-tuning PID control provides optimum thermal process
  • Adjustable high-limit OTP
  • LED display of actual temperature vs. setpoint
Light gauge overbend (More Product Information