Protektive Pak® Reel/Dip Tube Bin Boxes, Desco Industries
Supplier: Protektive Pak
37611 37130 37132 37134 37131 37133 37135
30000-196EA 62.36 CAD
30000-196 30000-864 30000-866 30000-868 30000-870 30000-872 30000-874
Protektive Pak® Reel/Dip Tube Bin Boxes, Desco Industries
Boxes ESD-Safe Boxes
Reusable boxes are designed to safely store both reels and rail/dip tubes in the same bin box
May be used for storing, kitting, or transporting during work-in-process operations. Made of impregnated, corrugated cardboard. A buried shielding layer minimizes sloughing and rub-off contamination. Feature a static-dissipative surface of 107–109ohms/sq. These reusable boxes are constructed with double sides and ends for durability. Made from 100% recycled material and are 100% recyclable. Cell identification labels are included. Boxes are shipped knocked-down; minimal assembly required.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable