Manual Pellet Press, Carver
Supplier: Carver
4350
29999-808EA 13873.38 CAD
29999-808
Manual Pellet Press, Carver
Presses
This compact 12-ton pellet press with 10.2cm (4") diameter platen is perfectly suited for KBr pellet pressing
It can also be used for glove box applications, compound and formula testing, compression strength tests, fluid extractions, and on-site field testing.
Ordering information: Press includes integral safety shield, buffer plate, 13mm evacuable die, and counter-bored platen for positive die alignment.