Corning® Storage Mats, Corning
Supplier: Corning
3080 3083 3346 3341
29445-122CS 715.64 CAD
29445-122 33500-570 80081-124 80081-114
Corning® Storage Mats, Corning
Seals Microplate Seals
Multiple formats for specific and precise fit on 96-well and 384-well plates and blocks.
  • Non sterile
  • Not treated
Round well microplate storage mat: Fits Corning® 96 round well plates and blocks.
Manufactured from ethyl vinyl acetate (EVA), compatible with DMSO
Certified free from DNase and RNase
Can be applied manually or with applicator
Square well storage block sealing mat: Fits Corning® 96 square 2 ml well storage blocks.
Manufactured from ethyl vinyl acetate (EVA), compatible with DMSO
Certified free from DNase and RNase
Can be applied manually orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable