Costar® Low Binding Microcentrifuge Tubes, Polypropylene, Corning
Supplier: Corning
3206
29442-578CS 114.17 CAD
29442-578
Costar® Low Binding Microcentrifuge Tubes, Polypropylene, Corning
Tubes Microcentrifuge Tubes
Low binding microcentrifuge tubes feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in better sample recovery.
With external graduations and frosted writing spot for easy sample identification. Tubes are disposable and autoclavable. Color: natural.
Certifications: Certified RNase- and DNase-free.