VWR® Blotting Paper, Grade 703

Supplier: VWR International
28298-020 28298-022 28298-024 28298-026 28298-028 28298-030 28298-020 28298-022 28298-026 28298-030 28298-024 28298-028
28298-020PK 777.91 CAD
28298-020 28298-022 28298-024 28298-026 28298-028 28298-030 CA28298-020 CA28298-022 CA28298-026 CA28298-030 CA28298-024 CA28298-028
VWR® Blotting Paper, Grade 703
Blotting Papers
Grade 703 blotting papers are ideal for use as wicks in protein and nucleic acid blotting.

  • Made of 100% cotton fiber
  • Uniform and smooth
  • Produced using ultrapure water
  • Contains no additives

Ideal for use as wicks in protein and nucleic acid blotting. Provides a uniform flow of buffer through the gel to the transfer membrane in a blotting sandwich. Also suitable for use in removing gels from glass supports.

Ordering information: Available cut to size, preventing the development of air pockets between the gel and transfer membrane. For moreMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR