Nalgene® Filter Holder with Funnel, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS0310-4050 DS0310-4000
28199-437CS 488.5 CAD
28199-437 28199-441
Nalgene® Filter Holder with Funnel, Thermo Scientific
Filter Holders
Polysulfone filter holder with funnel can be used with any filtering flask or manifold that accepts a suitable stopper with hole.
Note: For lab use only. Not for in vitro diagnosis or parenterals.
Autoclavable.
Ordering information: Unit comes complete with analytical filter support, sterilization filter support, and vacuum gasket, which takes the place of a No.7 or 8 rubber stopper with 14.3 mm (9/16") hole.
Order Now
SPECIFICATIONS
Filtration area 13.3 cm²
Environmentally Preferable