Nalgene® Rapid-Flow™ Sterilization Filter Units, Cellulose Nitrate Membrane, Thermo Scientific
Supplier: Thermo Fisher Scientific
125-0020 125-0045 125-0080 126-0045 126-0080 450-0080 450-0020 127-0020 127-0045 127-0080 126-0020 450-0045
28199-076CS 287.17 CAD
28199-076 28199-098 28199-101 28199-123 28199-134 28199-144 28199-155 28199-268 28199-279 28199-280 28199-112 28199-177
Nalgene® Rapid-Flow™ Sterilization Filter Units, Cellulose Nitrate Membrane, Thermo Scientific
Filters Bottle Top Filters
For easy, fast, sterile filtration of 150 to 1000 ml volumes.
  • Instantly ready for biological filtration, cold sterilization, or cleanup of particulated solutions or buffers
  • Components separate easily for pouring
  • No need to transfer sterile filtrate to another container
  • Graduations on uppers and receiver
  • Non-toxic, triton-free, preassembled and radiation sterilized
Nalgene 1¹/₂-turn threaded screw closure guarantees leakproof seal and eliminates pH shift in receiver bottles. Receiver bottles can be used to store filtrate. Filter cup is printed withMore Product Information