Nalgene® Filling Bell, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS0390-0070
28131-957CS 121.68 CAD
28131-957
Nalgene® Filling Bell, Polycarbonate, Thermo Scientific
Bottles Cell Culture Bottles
Transparent, autoclavable filling bell is for use in transferring sterile media aseptically from large containers to media bottles or culture vessels in microbiology and cell culture labs.
Autoclavable polypropylene tubing adapter accepts 6.3, 9.5, and 12.7 mm Int.Ø (¹/₄, ³/₈, ¹/₂") tubing.
Caution: For research use only; not for in vitro diagnosis or parenterals.