WHEATON® Filtration Assembly with PTFE Faced Support and No. 8 Stopper Connections, 47 mm, DWK Life Sciences
Supplier: DWK Life Sciences
419370
27589-118CS 831.89 CAD
27589-118
WHEATON® Filtration Assembly with PTFE Faced Support and No. 8 Stopper Connections, 47 mm, DWK Life Sciences
Funnels Filtration Funnels Filtration Apparatus Glass
The filtration assembly with fritted glass support is designed to handle up to 1000 ml of liquid for analysis of particulate or microbiological contamination.
  • Glass components manufactured from WHEATON 33 low extractable borosilicate glass
  • Recommended for autoclaving with the filter in place
  • PTFE coating prevents the filter from adhering to the ground glass surfaces
Flasks have a #2 hose connection for 6mm (1/4”) I.D. tubing. Flasks are not included.
Certifications: Glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable