Owl™ Aluminum Backed Sequencer System, Models S1S, S2S, S3S and S4S, Thermo Scientific
Supplier: Thermo Fisher Scientific
S3S S4S
27372-788EA 3934.2 CAD
27372-788 27372-796
Owl™ Aluminum Backed Sequencer System, Models S1S, S2S, S3S and S4S, Thermo Scientific
Electrophoresis Sequencers
Design provides flat, even banding patterns across all lanes and shortens pre-run times.
The upper buffer chamber (UBC) uses an in-line stopcock to drain into a removable container on the back of the unit
Integral side clamps ensure a leakproof seal and even pressure across the gel. Sliding side clamps are mounted on the unit for easy set-up.
Floating, anodized aluminum heat sink moves with the unit as it heats and expands, reducing glass breakage. Heat sink holds plates tightly, providing effective heat dissipation.
A silicone gasket provides a leakproof seal at the UBC. TheMore Product Information