Glass Plates for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Supplier: Thermo Fisher Scientific
ADJ1-38G ADJ1-40G ADJ1-40R P1-14G P1-14R P2-20FG P2-20FR ADJ1-23G P5-16R P10-20R
27372-014EA 53.15 CAD
27372-014 27372-016 27372-020 27372-516 27372-522 27372-524 27372-580 27372-582 27373-309 27373-428 27373-441
Glass Plates for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Electrophoresis Plates and Sheets
Plates are available in three styles: notched glass, blank glass, and offset glass.
Heat sinks are recommended when running sequencing gels.