PYREX® Folin-Wu Digestion Tube, Corning
Supplier: Corning
7900-25
26658-040CS 58.25 CAD
26658-040
PYREX® Folin-Wu Digestion Tube, Corning
Tubes Digestion Tubes
Made of PYREX® borosilicate glass.
  • Round bottom
  • With engraved white-filled graduations at 25 and 50 ml
  • Tolerance of ±0.4 ml
The Folin-Wu digestion tubes are graduated with a beaded rim and are fabricated from specially selected tubing for product durability.