PYREX® Nitrogen Distilling Apparatus, Corning
Supplier: Corning
3340-500
26315-007CS 1074.7 CAD
26315-007
PYREX® Nitrogen Distilling Apparatus, Corning
Distillation Components Distillation Sets
Distilling apparatus to determine nitrogen levels in organic compounds.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The assembly includes a Kjeldahl flask specifically developed for this process and a Graham condenser with 10 mm O.D. (3/8") tubulations. Both the digestion and distillation stages can be performed in the flask. The flask has a [ST] 29/42 outer joint. The condenser has a [ST] 19/38 outer joint. Height: 41 cm (161/8"). Width: 30 cm (1113/16").More Product Information