Barnstead™ B-pure™ Half-Size Filter Holder, Thermo Scientific
Supplier: Thermo Fisher Scientific
D5839
26305-400EA 1109.32 CAD
26305-400
Barnstead™ B-pure™ Half-Size Filter Holder, Thermo Scientific
Filter Holders
These filter holders are used with Barnstead™ B-Pure™ 1/2 size filters to pre-treat feed water or for a final filter to help ensure consistently pure water.