Spectra/Por® 7 Dialysis Membranes, MWCO 1000 to 50000
Supplier: Spectrum Laboratories
132103 132104 132105 132107 132108 132109 132110 132111 132112 132125 132126 132127 132128 132129 132130 132113 132114 132115 132118 132119 132120 132121 132122 132123 132124 132131 132116 132117 132104T 132105T 132107T 132108T 132111T 132112T 132126T 132127T 132129T 132115T 132119T 132120T 132130T
25223-050EA 869.73 CAD
25223-050 25223-071 25223-093 25223-130 25223-151 25223-173 25223-195 25223-210 25223-231 25223-490 25223-515 25223-537 25223-559 25223-570 25223-592 25223-650 25223-672 25223-694 25223-785 25223-800 25223-821 25223-887 25223-901 25223-923 25223-945 28170-170 25223-719 25223-763 10768-600 10768-602 10768-604 10768-606 10768-608 10768-610 10768-612 10768-614 10768-616 10768-618 10768-620 10768-622 10768-624 10768-682 10768-684 10768-686 10768-688 10768-690 10768-692 10768-694 10768-696 10769-058 10769-060 10769-062 10769-064 10769-066
Spectra/Por® 7 Dialysis Membranes, MWCO 1000 to 50000
Tubing Dialysis Tubing
Free of trace metal and sulfur for separations involving a possible sensitivity to such materials.
  • Transparent and flexible
  • Minimum sulphur and heavy metal contamination
Supplied wet in 0.1% sodium azide as a preserving agent. Just rinse in distilled water and condition in dialyzing buffer and membrane is ready for use.
Roll length: 5 m (16.4')
Order Now
SPECIFICATIONS
Environmentally Preferable