Nalgene® Graduated Cylinders, PMP, Thermo Scientific
Supplier: Thermo Fisher Scientific
3663-0010 3663-0025 3663-0050 3663-0100 3663-0250 3663-0500 3663-1000 3663-2000 3663-4000
24780-028CS 1000.85 CAD
24780-028 24780-040 24780-061 24780-083 24780-108 24780-120 24780-141 24780-163 24780-185
Nalgene® Graduated Cylinders, PMP, Thermo Scientific
Cylinders Measuring Cylinders
Graduated cylinders are suitable for food and beverage use.
  • Calibrated to contain/deliver
  • Perfect transparency
  • Remarkable heat resistance
  • Excellent chemical resistance
  • Shatter-proof
  • Withstands continuous usage at 350°F
Molded-in graduation lines on the back of the cylinders ensure easier, more accurate fluid level readings. Large blue base resists tipping and is molded of superior strength polypropylene that resists cracking, chipping, and breakage.
Certifications: Cylinders meet ASTM Class B E1272 and ISO Standard 6706. Polymethylpentene resinMore Product Information