Nalgene® PMP Hydrometer Jar, Thermo Scientific
Supplier: Thermo Fisher Scientific
6230-0500
24661-255CS 440.2 CAD
24661-255
Nalgene® PMP Hydrometer Jar, Thermo Scientific
Cylinders Hydrometer Cylinders
Hydrometer jar with cylinder of crystal-clear polymethylpentene (PMP)
Broad plastic base provides good stability. Suitable for use as lactometer cylinder.
Superior chemical resistance and safety. Withstands continuous usage at up to 135°C. Autoclavable. Tube ungraduated and without identification. No overflow cup required for reading.