PYREX® Low Actinic Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
Supplier: Corning
52300-650
23127-022CS 257.07 CAD
23127-022
PYREX® Low Actinic Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
Condensers
This PYREX® air condenser is made from low actinic glass.
  • Air Reflux style condenser
  • Has an inner 24/40 standard taper joint with a drip tip
To protect light sensitive materials and is useful for condensing materials with boiling points above 150 °C (302 °F).