Hog Bristle and Plastic Handle Sash Brush, Gordon Brush
Supplier: Gordon Brush
900162
22940-750EA 68.16 CAD
22940-750
Hog Bristle and Plastic Handle Sash Brush, Gordon Brush
Brushes
This Size 6 hog bristle and plastic handle sash brush has an oval shape, and is excellent for dusting and removing debris.
  • Features a metal ferrule
Large tapered plastic handle.
Order Now
SPECIFICATIONS
Environmentally Preferable