Aluminum Handle Brushes, Gordon Brush
Supplier: Gordon Brush
11CKAG-12 11HHAG-12 33CKAG-12 33HHAG-12
22940-548EA 110.82 CAD
22940-548 22940-550 22940-552 22940-554
Aluminum Handle Brushes, Gordon Brush
Brushes
Antistatic brush is ideal for fine detailing and use in tight areas
Fibers are hand laced for better fiber retention and durability.
Order Now
SPECIFICATIONS
Environmentally Preferable