Corning® Spinner Flasks with Angled Sidearms, Corning
Supplier: Corning
4500-500 4500-1L 4500-250 4500-3L 4500-6L 4500-8L 4500-15L 4500-36L 4502-3L 4502-8L 4502-15L 4502-36L
22877-076CS 1350.69 CAD
22877-076 22877-078 22877-074 22877-080 22877-082 22877-084 22877-086 22877-088 22877-090 22877-094 22877-096 22877-098
Corning® Spinner Flasks with Angled Sidearms, Corning
Flasks Spinner Flasks
Borosilicate glass with side arms, non sterile.
  • Baffles enhance aeration and agitation of flask contents
  • Impeller design ensures optimal stirring
  • Complete with impeller and caps
  • Side arm design permit easy access of 25 and 50 ml pipetes
Order Now
SPECIFICATIONS
Environmentally Preferable