Tweezertrodes™ Kits, BTX™
Supplier: Harvard Apparatus
45-0165 45-0166
22221-518EA 3814.33 CAD
22221-518 22221-520
Tweezertrodes™ Kits, BTX™
Electrodes Electroporation Electrodes
Tweezertrodes™ are reusable, non-invasive, tweezer-style in vivo and in utero electrodes for drug or gene delivery in animal tissues
Tweezertrodes™ consist of a standard 11.5cm tweezer that has been modified with stainless steel circular or disk electrodes at the tip. The gap between electrode tips can be adjusted from under 1mm to over 2cm. Tweezertrodes™ are available in various sizes and two different metal alloys (Platinum or Stainless Steel). They may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide. These electrodes connect to BTX™ generators usingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Voltage range 0 V - 1 kV
Pulse length range 10 µs to 99 ms in PBS (Phosphate Buffer Saline)