VWR® Premoistened Cleanroom Wipers
Supplier: VWR International
21910-009 21910-110 21910-111
21910-009PK 43.18 CAD
21910-009 21910-110 21910-111
VWR® Premoistened Cleanroom Wipers
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Premoistened wipers are designed for environmentally sensitive areas, and are ideal for wiping optical parts, delicate glassware, electrodes, cuvettes, microscopes, and lenses.
  • Strong, non-woven cellulose/polyester blend
  • Soft and lint-free
  • Premoistened or dry
  • For use in environmentally sensitive applications
Wipers are available premoistened with 70% reagent-grade isopropyl alcohol and 30% reagent-grade deionized water, 100% pure reagent-grade deionized water, or an eyeglass cleaning solution.
Strong, nonwoven cellulose/polyester wipers are soft,More Product Information