Tergazyme® Enzyme-Active Powder Detergents
Supplier: Alconox
1325 1301 1303 1350 1303 1304
21837-151EA 710.42 CAD
21837-151 21837-162 21837-164 21837-166 CA11005-586 21837-118
Tergazyme® Enzyme-Active Powder Detergents
Cleaning Agents Detergents and Surface Cleansers
Tergazyme is a concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning. Excellent for removal of proteinaceous soils, tissue, blood and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass with no interfering residues. Ideal as a cleaning agent in Reverse Osmosis and Ultra-Filtration Systems.
  • Biodegradable and readily disposable
  • Replaces corrosive acids and hazardous solvents
  • Protease enzyme removes proteinaceous soils, tissue, blood and body fluids
  • Free rinsing to give you reliable results
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable