Alconox® Powdered Precision Cleaners
Supplier: Alconox
1125 1150 1101 1103 1104 1112
21835-054EA 782.84 CAD
21835-054 21835-065 21835-087 21835-089 21835-032 21835-123
Alconox® Powdered Precision Cleaners
Cleaning Agents Detergents and Surface Cleansers
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass.
  • Biodegradable and readily disposable
  • Penetrating wetting power to save you time
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100
It can be Used to clean healthcare instruments,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable