Alcojet® Low-Foaming Powdered Detergents
Supplier: Alconox
1401 1403 1425 1450 1404
21834-045EA 2277.62 CAD
21834-045 21834-055 21834-075 21834-084 21834-051
Alcojet® Low-Foaming Powdered Detergents
Cleaning Agents Detergents and Surface Cleansers
Non-ionic, low foaming powder cleaner for use in labware washers, parts washers, power-spray systems, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues.
  • Concentrated
  • Biodegradable and readily disposable
  • High performance from your cleaning machinery without damaging washer parts
  • Free rinsing to give you reliable results and no interfering residues
  • Dilute 1:200
For removing soil, grit, grime,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
pH 12.0