Staticide® Acrylic Static-Dissipative Floor Finish, ACL Staticide
Supplier: ACL Staticide
40002 40005 40001
21831-011EA 6269.52 CAD
21831-011 21831-012 21831-009
Staticide® Acrylic Static-Dissipative Floor Finish, ACL Staticide
Surface Treatments Floor Treatments
Suitable for electronic production lines, computer rooms, and other high-tech static-sensitive environments
  • Tough, easy-to-use, nonmigratory acrylic copolymer is high gloss, slip- and detergent-resistant
  • Can be spray-buffed for easy maintenance
  • Can be machine-buffed and may be cleaned without loss of gloss on finish
For best results in curing, Staticide® neutralizer/cleaner should be used before applying finish (see Staticide® Antistatic Neutralizer/Cleaner, ACL series).
Charge generation <100 V
Static charge decay: 10 s
Surface resistance: –108More Product Information