VWR® Traceable® Dissolved Oxygen Meter
Supplier: VWR International
93676-35
21800-022EA 2071.96 CAD
21800-022
VWR® Traceable® Dissolved Oxygen Meter
Electrochemistry Meters Oxygen Meters
This dissolved oxygen meter simultaneously displays oxygen concentration and temperature.
  • Jumbo-size digits
  • Memory recall of readings
  • Can freeze display to capture readings
  • Computer output capabilities
Computer output allows it to be connected to a computer or data logger for monitoring and keeping results. Jumbo-size digits are 13/8" high. At the touch of a button the instrument recalls highest, lowest, and average readings. A Data Hold button freezes the display to capture readings.
Certifications: To ensure accuracy, an individually serial-More Product Information