Upchurch Scientific® Stainless Steel Frits, IDEX Health & Science
Supplier: Upchurch Scientific
C-128-31 C-425 C-420 A-418 A-103 A-420 A-337 A-100 A-331
21001-324EA 22.82 CAD
21001-324 53501-342 53501-346 21001-308 21511-428 53501-234 53501-486 21511-429 21511-453
Upchurch Scientific® Stainless Steel Frits, IDEX Health & Science
Filters Precolumn Filters and Frits
Upchurch Scientific® stainless steel frits are available in several porosities: 0.5, 2, 10, and 20 µm.
While all Upchurch Scientific® stainless steel frits are capable of withstanding pressures >5000 psi (>345 bar), the actual pressure holding capacity of each frit is usually dependent on the filter body in which it is placed.
Upchurch Scientific® Frit-In-A-Ferrule™ seal and filter simultaneously and can filter at any point in your system where ¹/₁₆ or ¹/₈" Ext.Ø tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. For more information and a listing of these products,More Product Information