Upchurch Scientific® Inlet Solvent Filters, IDEX Health & Science
Supplier: Upchurch Scientific
A-427 A-445 A-438 A-520X A-550 A-551 A-201 A-210 A-446 A-437 A-522X A-302A A-310 A-311A A-227A
21511-023EA 152.77 CAD
21511-023 53500-756 21001-092 21001-172 21001-174 21001-178 21001-180 53498-554 53498-556 21001-094 21001-166 21001-168 21001-170 21001-176 21511-006 21511-008 21511-010 53501-370 53501-418 97055-068
Upchurch Scientific® Inlet Solvent Filters, IDEX Health & Science
Filters Liquid Process Filters
It is good practice to filter solvents to prevent pump and other system component damage.
Upchurch Scientific® Inlet Solvent Filters are available in 2, 10, and 20 µm porosities for general use, preparative, and specialized applications. Large surface areas prevent pump cavitation while stainless steel and polymer versions offer chemical compatibility. Special 'Bottom-of-the-Bottle' versions are also available to allow solvent to be drawn to within ¹/₈" (or less) of the bottom of a solvent bottle. For inlet solvent filtering, 10 to 20 µm porosity is usually recommended to filter theMore Product Information