OPTI-LOK™ High Pressure Fittings
Supplier: Optimize
10-20-00272 10-20-00274 10-20-00276 10-20-00255 10-20-00256 10-20-00266 10-20-00251 10-20-00253 10-20-00258 10-20-00259 10-20-00263 10-20-00264 10-20-00265 10-20-00268 10-20-00269 10-20-00270 10-20-00271 10-20-00273 10-20-00275 10-20-00518 10-24-00367 10-24-00369 10-24-00370 10-24-00371 10-24-00373 10-24-00375 10-24-00376 10-20-00261
21510-159PK 655.79 CAD
21510-159 21510-163 21510-167 21510-680 21510-682 21510-696 77100-154 77100-156 77100-158 77100-160 77100-166 77100-168 77100-170 77100-172 77100-174 77100-176 77100-178 77100-180 77100-182 77100-188 77100-260 77100-264 77100-266 77100-268 77100-270 77100-272 77100-274 89522-310
OPTI-LOK™ High Pressure Fittings
Fittings and Connectors
OPTI-LOK Fittings are ideal for high-pressure connections with ¹/₁₆" outer Ø tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6000 psi.
  • Precision machined for optimal quality and thread consistency
  • Zero-dead-volume connections every time
  • Trouble-free operation at high pressures
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable