HC-75 Calcium Form Cation Exchange HPLC Columns, Hamilton Company
Supplier: Hamilton
79431 79436
21493-003EA 1440.11 CAD
21493-003 21493-006
HC-75 Calcium Form Cation Exchange HPLC Columns, Hamilton Company
Chromatography Columns
Hamilton Company offers one of the most comprehensive selections of chromatography columns in the industry.
This product employs polymeric cross-linked soft-gel materials for cation, ligand exchange separation of carbohydrates. Benefits and Features:
- Separate Mono and Disaccharides
- Separate oligomers from DP1 to DP8
- Compatible with RI detectors
- 9 µm particle size
- Multiple column internal diameters available
- Stainless Steel or PEEK hardware available
HC-75 gel-type cation exchange columns are especially useful for determining ethanol in fermentation broths and for separatingMore Product Information