UVP Chromato-Vue® Viewing Cabinets, Analytik Jena
Supplier: Analytik Jena US
95-0257-01 95-0257-02 95-0253-01 95-0253-02 95-0072-01 95-0072-07 95-0072-06 95-0020-04 95-0020-05 95-0022-01 95-0022-02
21474-406EA 5884.6 CAD
21474-406 21474-408 21474-608 21474-610 21475-017 21475-040 21475-039 21475-061 21475-083 21475-094 21475-108
UVP Chromato-Vue® Viewing Cabinets, Analytik Jena
Lights UV Viewing Cabinets
UVP Chromato-Vue Viewing Cabinets provide a darkroom environment for viewing materials, non-destructive testing (NDT), inspection, quality control and chromatography applications. All models include an integrated, contrast control filter to protect the eyes from harmful shortwave radiation and blocks the 'blue haze' associated with longwave UV. Viewport has adequate space for eyeglasses. A lightweight access curtain allows for easy entry and blocks out external light. A variety of cabinets are available to choose from.
C-75, C70 and C71, models are designed for the highest performance andMore Product Information