G-Tube® Microcentrifuge Tubes, 2.0 ml, Bio Plas
Supplier: Bio Plas
4057 4050 4051 4052 4053 4054 4056 4059
12777-148CS 871.26 CAD
12777-148 21150-486 21150-488 21150-490 21150-492 21150-494 21150-498 21150-502
G-Tube® Microcentrifuge Tubes, 2.0 ml, Bio Plas
Tubes Microcentrifuge Tubes
Bio Plas G-Tube® is built to withstand speeds over 30000×g.
  • Made of polypropylene
  • Non-pyrogenic and endotoxin-free
  • RNase- and DNase-free
Flat top lid forms an extra tight seal that will not come open during processing. G-Tubes are manufactured from medical grade resins and feature textured writing surfaces on both the lid and sides. These low retention, autoclavable tubes can withstand temperature ranges from –80 to 121 °C (–112 to 250 °F).
Order Now
SPECIFICATIONS
Environmentally Preferable