Microcentrifuge Tubes with Snap Cap, Bio Plas
Supplier: Bio Plas
4003 4004 4005 4025 4151 4152 4153 4154 4156 4157 4158
21150-326CS 1112.81 CAD
21150-326 21150-328 21150-330 21150-332 21150-336 21150-338 21150-340 21150-342 21150-346 21150-348 21150-350 21150-352
Microcentrifuge Tubes with Snap Cap, Bio Plas
Tubes Microcentrifuge Tubes
The Snap-Cap Microcentrifuge Tube gives a firm, dependable seal, and its tough virgin-polypropylene construction withstands the rugged G-forces of high-speed centrifugation.
  • Ultra low retention
  • Snap-Caps for optimal sealing
  • RNase and DNase free and non-pyrogenic
Used for specimen collection, mixing, electrophoresis sampling/handling, enzyme assays, radioassays, immumoassays and many other applications. The Snap-Cap Microcentrifuge Tube’s smooth, conical interior lets remove entire sample with ease, and without material hangup. Fits Beckman™, Eppendorf™More Product Information