96-Place Reversible Microcentrifuge Tube Racks, Bio Plas
Supplier: Bio Plas
0090 0091 0092 0093 0094 0095 0096 0097 0099
21150-228CS 896 CAD
21150-228 21150-230 21150-232 21150-234 21150-236 21150-238 21150-240 21150-242 21150-244
96-Place Reversible Microcentrifuge Tube Racks, Bio Plas
Racks Tube Racks
These 96-place reversible racks are intended for use with 0.5mL, 1.5mL, and 2.0mL snap cap and screw cap microcentrifuge tubes.
  • Racks are autoclavable
  • Two frosted writing areas are provided for sample identification
A clear polypropylene cover easily snaps on to keep samples firmly in place during transportation and storage. A recessed area in the top of the covers allows for secure stacking of racks atop one another. The snap on cover is designed to accommodate the extra height of Bio Plas Screw Cap Microcentrifuge tubes.
Ordering information: More Product Information