PYREX® Conical Centrifuge Tube with Beaded Rim, Corning
Supplier: Corning
8060-15 8060-50
21021-188CS 285.79 CAD
21021-188 21021-202
PYREX® Conical Centrifuge Tube with Beaded Rim, Corning
Tubes Centrifuge Tubes
Made of PYREX® borosilicate glass, conical, beaded.
  • Wide neck
  • Excellent chemical resistance
  • Excellent temperature resistance
These centrifuge tubes has uniform glass distribution in the wall, taper and bottom provides improved mechanical strength, prevents the formation of failure-causing stress regions during centrifuging and insures the highest degree of accuracy.