Distillation Glassware Set with Wheaton Connection®, Semi-Micro Kit™, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
165996
20762-096CS 150.44 CAD
20762-096
Distillation Glassware Set with Wheaton Connection®, Semi-Micro Kit™, Wheaton
Distillation Components Distillation Sets
Adapter has an “O” ring that holds a thermometer or gas inlet tube securely.
A grease-free WHEATON Connection® on one end provides an inert connection with external-threaded glass components. Adapter can also be used with gas inlet tubing and will accommodate any tube with an OD of 5 to 7 mm.