Laboratory Pipetting Needles with 90° Blunt Ends, Cadence Science®
Supplier: Cadence Science
7930 7931 7934 7935 7936 7937 7938 7939 7941 7942 7956 7954 7950 7952 7932 7933 7940 7951 7953 7955 7957 9870 9871 9873 9874 9875 9876 9872
20068-672PK 721.03 CAD
20068-672 20068-674 20068-676 20068-678 20068-680 20068-682 20068-684 20068-686 20068-688 20068-690 20068-692 20068-694 20068-696 20068-698 20068-779 20068-781 20068-783 20068-785 20068-787 20068-789 20068-791 20069-028 20069-030 20069-034 20069-036 20069-038 20069-040 89234-232
Laboratory Pipetting Needles with 90° Blunt Ends, Cadence Science®
Syringe Needles
Cadence Science® pipetting needles are available in a wide choice of sizes for use in laboratory procedures.
Nonsterile. Packaged 12 needles per box.
These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors. Pipetting needles are available with three distinct hub styles; standard nickel plated hubs with 304 stainless steel cannulae for regular applications, 304 Stainless steel hubs with 304 stainless steel cannulae for special applications with corrosive materials or critical procedures, or 316 Stainless Steel Micro-Mate®More Product Information