Black Max® 380™ Toughened Instant Adhesive, Loctite
Supplier: Loctite
135423
19807-526CS 124.98 CAD
19807-526 19807-528
Black Max® 380™ Toughened Instant Adhesive, Loctite
Adhesives
Black Max® 380™ is the original elastomer-modified instant adhesive.
  • Adhesive cures quickly at room temperature
  • Fills gaps up to 0.015 cm (0.006")
  • Excellent peel, impact, and shear strength
Adhesive bonds metal, rubber, and plastics, and resists peeling, cleavage loading, and stresses caused by thermal shock.