SP Bel-Art Burettes, Automatic Self-Zeroing, Class B, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37918-0110 H37918-0109 H37918-0111 H37918-0112
13197-436EA 899.87 CAD
13197-436 17579-427 17579-482 17579-518
SP Bel-Art Burettes, Automatic Self-Zeroing, Class B, Bel-Art Products, a part of SP
Burettes
These self-zeroing glass burettes have easy-to-read blue graduations against a white background.
  • Easy-to-read measurements
  • Blue graduations on white background
  • Polyethylene construction
  • Class B graduation accuracy
The delivery end is fitted into a plastic push button stopcock which permits drop by drop delivery
A polyethylene filling tube extends from the top of the burette into the bottle reservoir. Bottle, screw cap, and round base are polyethylene plastic. Graduation accuracy is Class B.
A small screw clamp adjusts drop speed as needed forMore Product Information