KIMAX® Micro Burets, Serialized and Certified, Class A, Three-Way PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17115F-5 17115F-10
17492-103CS 1129.16 CAD
17492-103 17492-147
KIMAX® Micro Burets, Serialized and Certified, Class A, Three-Way PTFE Stopcock, Kimble Chase
Burettes
Three-way PTFE [PS] stopcock, plain top and stopcock fill tube
Designed to satisfy minimum delivery requirements for ASTM E287 Class A using equipment which is individually calibrated and certified against equipment whose calibration is traceable to the National Institute of Standards and Technology. Each buret is permanently marked with an individual serial number. Stopcocks are designed to comply with ASTM E911.
Constructed to minimize fouling by dirt or grease. The tip should be cleaned as part of a regular maintenance program (cleaning wire included).
Filling tube accepts 0.25"More Product Information