HyClone™ Fetal Bovine Sera Defined (United States), HyClone Products (Cytiva)
Supplier: Cytiva
SH30070.03HI SH30070.03 SH30070.03
16777-530EA 2111.95 CAD
16777-530 16777-006 CA16777-006 CA16777-530 CA16750-030 CA16777-002 CA82013-568
HyClone™ Fetal Bovine Sera Defined (United States), HyClone Products (Cytiva)
Sera (Serum)
Defined fetal bovine serum (FBS) is the highest quality of FBS available and is very widely used by those cell culturist who have a concern for viral contaminants and require an extensive biochemical profile. Defined FBS is filtered through multiple 40 nm (0.04 µm) pore-size related filters, which are the most retentive filters used in commercial FBS production. Over 50 components are analysed and the results are included on the biochemical assay.
  • Filtered through serial 40 nm pore-size rated filters
  • Maximize cell culture growth
  • U.S. sourced