VWR® Wide Mouth Safety Wash Bottles, LDPE
Supplier: VWR International
16649-900 16649-901 16649-905 16649-915 16649-917 16649-918 16649-925 16649-940 16649-943 16649-950 16649-955 16649-966 16649-910 16649-911 16649-935 16649-945 89141-042 89141-046
16649-900PK 125.69 CAD
16649-900 16649-901 16649-905 16649-915 16649-917 16649-918 16649-925 16649-940 16649-943 16649-950 16649-955 16649-966 16649-910 16649-911 16649-935 16649-945 89141-042 89141-046
VWR® Wide Mouth Safety Wash Bottles, LDPE
Bottles Wash Bottles
Easy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information.
  • Easy-to-read, low-density polyethylene dispensing bottles are break-resistant
  • Intended for short-term dispensing use
  • Include DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ information
Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label.
Toluene bottle is solid red with whiteMore Product Information