Hazardous Materials Shippers, Safety-Coated Glass, UN Marked, Qorpak®
Supplier: Berlin Packaging
HMS-6653M HMS-6654M HMS-6656M HMS-6651M HMS-6645M HMS-6644M HMS-6642M HMS-6643M
16465-001CS 640.55 CAD
16465-001 16465-003 16465-005 16465-007 16465-009 16465-011 16465-013 16465-015
Hazardous Materials Shippers, Safety-Coated Glass, UN Marked, Qorpak®
Mailers and Shippers
Individual convenience packs suitable for shipping hazardous materials.
  • Plastic coated narrow-mouth bottles for liquids
  • Plastic coated wide-mouth bottles for solid and viscous materials
  • 120mL shippers are not plastic coated
Certified packaging is supplied complete with safety-coated glass container, Thermoset closure with foam-backed fluoropolymer resin liner, corrugated outer packaging, cushioning material, absorbent, and tape.
Meet UN 4GV requirements (labels required).
Warning: Shipper must not make any substitutions. Failure to heed thisMore Product Information