PYREX® Dropping Bottle with Bulb and Pipette, Corning
Supplier: Corning
1340-125
16359-060CS 1923.73 CAD
16359-060
PYREX® Dropping Bottle with Bulb and Pipette, Corning
Bottles Dropping Bottles
Narrow mouth droppings bottles, made of PYREX® borosilicate glass.
  • Reusable bottle
  • Pipette with suction bulb
  • Hollow glass stopper style
Recommended for use where preventing contamination by solubility or pH change is important.