Laboratory Bottles, Wide Mouth, HDPE, Bulk Packed, Dynalon
Supplier: Dynalon
301615-0004 301615-0008 301615-0016 301615-0064
16354-814CS 1557.85 CAD
16354-814 16354-816 16354-818 30620-172
Laboratory Bottles, Wide Mouth, HDPE, Bulk Packed, Dynalon
Bottles Plastic Bottles
Leakproof, rigid, and translucent, these high density polyethylene wide mouth lab bottles offer excellent chemical resistance.
Supplied with a polypropylene closure, they are ideal for storage and handling of liquids or solids. Manufactured from FDA approved resin for non-contaminating storage of chemicals and food grade products. Bulk packed with closures included but not attached.