Standard Wide Mouth Bottles, Clear, Qorpak
Supplier: Qorpak
GLC-01754 GLC-01750 GLC-01779 GLC-01843 GLC-01870 GLC-01889 GLC-01775 GLC-01848 GLC-01872 GLA-00869 GLA-00871 GLA-00875 GLA-00882 GLA-00883 GLC-01770 GLC-01800 GLC-01858 GLC-01882 GLC-01827 GLC-01877 GLC-01762 GLC-01792 GLC-01737 GLC-01833 GLC-01755 GLC-01743 GLC-01744 GLC-01739 GLC-01738 246341 261438 242600 272013 272012 GLC-01852
11311-554CS 117.9 CAD
11311-554 16054-168 16054-170 16054-206 16081-043 16081-045 16081-050 16085-030 16085-230 16085-232 16085-234 16085-236 16087-306 16152-168 16193-388 16193-402 16193-526 16193-548 16193-560 16193-606 16193-810 16193-843 16193-876 16194-500 16194-504 16195-780 16195-805 16195-827 16195-830 16195-860 16195-871 16196-207 16196-229 16196-262 16196-284 18087-090 36319-155 36319-195 36319-199 36319-219 36319-225 36319-226 36319-229 36319-248 89176-848 89176-850 93001-560 16196-240 89204-684 89204-686 89204-688 89204-690 89204-692 89204-694 10053-746 10798-936 10022-242 76170-360 76170-362 76170-366 76170-414 76419-806 76419-808 76419-810 76419-812 76419-814 76487-214
Standard Wide Mouth Bottles, Clear, Qorpak
Bottles Glass Bottles
Clear, flint glass standard wide mouth bottles feature a wide opening making them the ideal general-purpose bottle.
Applications include pharmaceutical, chemical, environmental, food packaging and food storage.
Ordering information: Bottles 36319-155, 36319-195, 36319-199, 36319-219, 36319-225, and 89204-692 are supplied pre-cleaned for volatiles. Bottles 89176-848, 89176-850, 93001-560, 93001-562, 16054-206, 76487-214 and 10053-746 are supplied pre-cleaned and certified for semi-volatiles.