PYREX® Media/Storage Bottles, Narrow Mouth, Round, with Screw Cap, Corning
Supplier: Corning
1395-50 1395-25 1395-5L 1395-10L 1395-100 1395-250 1395-500 1395-1L 1395-2L 1395-150 1395-750 1395-3X
16157-068CS 262.11 CAD
16157-068 16157-071 16157-250 16157-282 16157-103 16157-136 16157-169 16157-191 16157-227 89427-656 89427-658 89427-655
PYREX® Media/Storage Bottles, Narrow Mouth, Round, with Screw Cap, Corning
Bottles Glass Bottles
Heavy duty bottles, made of PYREX® borosilicate glass, with orange PP linerless, one-piece autoclavable plug seal screw with a drip-free pouring ring.
  • Permanent white enamel graduations with marking spots
  • Glass bead indicates full capacity line
  • Very good temperature resistance, very good chemical resistance
Graduated, are used for storage as well as mixing and sampling.
Further colour-coded PP caps, PBT caps, ETFE pouring rings (for dry heat sterilization (180 °C)), and vented membrane caps are available.
Caution: Do not tighten cap immediatelyMore Product Information